accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH3O

CAS RN 105-46-4

IUPAC Name: butan-2-yl acetate
Synonyms: sec-butyl acetate S-butylacetate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: DCKVNWZUADLDEH-UHFFFAOYNA-N
SMILES: CCC(C)OC(C)=O
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)