accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 105-53-3

IUPAC Name: 1,3-diethyl propanedioate
Synonyms: diethyl malonate
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: IYXGSMUGOJNHAZ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)OCC
Boiling Point 
  • (3)
  • (2)
  • (4)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (9)
Percent Purity 
  • (2)
  • (7)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (2)
  • (8)
  • (3)
  • (2)
  • (4)
  • (5)
  • (5)
  • (9)
  • (2)
  • (7)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (10)