accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 105-54-4

IUPAC Name: ethyl butanoate
Synonyms: ethyl butyrate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: OBNCKNCVKJNDBV-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (8)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)