accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-60-2

IUPAC Name: azepan-2-one
Synonyms: caprolactam epsilon-caprolactam 1,6-hexolactam 2-oxohexamethylenimine e-kaprolaktam 6-caprolactam 6-hexanelactam 1,6-hexalactam caprolactam (substance) lactam, aminocaproic
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: JBKVHLHDHHXQEQ-UHFFFAOYSA-N
SMILES: O=C1CCCCCN1
Boiling Point 
  • (3)
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (6)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (7)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (6)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)