accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 105-66-8

IUPAC Name: propyl butanoate
Synonyms: propyl butyrate N-propylbutyrate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: HUAZGNHGCJGYNP-UHFFFAOYSA-N
SMILES: CCCOC(=O)CCC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results