accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 105-74-8

IUPAC Name: dodecanoyl dodecaneperoxoate
Synonyms: peroxide, bis(1-oxododecyl)
Molecular Weight (g/mol): 398.63
Molecular Formula: C24H46O4
InChi Key: YIVJZNGAASQVEM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)