accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-81-7

IUPAC Name: 1-(2-hydroxyethyl)-3-(prop-2-en-1-yl)thiourea
Synonyms: 1-(2-hydroxyethyl)-3-(prop-2-en-1-yl)thiourea
Molecular Weight (g/mol): 160.24
Molecular Formula: C6H12N2OS
InChi Key: VUVPNTYTOUGMDG-UHFFFAOYSA-N
SMILES: OCCNC(=S)NCC=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)