CAS RN 105-86-2
IUPAC Name:
(2E)-3,7-dimethylocta-2,6-dien-1-yl formate
Synonyms:
geranyl formate geranyl formate (natural)
Molecular Weight (g/mol):
182.26
Molecular Formula:
C11H18O2
InChi Key:
FQMZVFJYMPNUCT-YRNVUSSQSA-N
SMILES:
CC(C)=CCC\C(C)=C\COC=O
1
–
1
of
1
results
1
Geranyl Formate 55.0+%, TCI America™
CAS: 105-86-2 Molecular Formula: C11H18O2 Molecular Weight (g/mol): 182.263 MDL Number: MFCD00021047 InChI Key: FQMZVFJYMPNUCT-YRNVUSSQSA-N PubChem CID: 5282109 ChEBI: CHEBI:31648 IUPAC Name: [(2E)-3,7-dimethylocta-2,6-dienyl] formate SMILES: CC(=CCCC(=CCOC=O)C)C