CAS RN 10500-11-5
CAS RN 10500-11-5
IUPAC Name:
pent-1-en-3-yl acetate
Synonyms:
pent-1-en-3-yl acetate
Molecular Weight (g/mol):
128.17
Molecular Formula:
C7H12O2
InChi Key:
MRLKTTBPWZXARX-UHFFFAOYNA-N
SMILES:
CCC(OC(C)=O)C=C
1
–
1
of
1
results
1
1-Penten-3-yl Acetate 97.0+%, TCI America™

CAS: 10500-11-5 Molecular Formula: C7H12O2 Molecular Weight (g/mol): 128.171 MDL Number: MFCD02093427 InChI Key: MRLKTTBPWZXARX-UHFFFAOYSA-N Synonym: Acetic Acid 1-Ethylallyl Ester, Acetic Acid 1-Penten-3-yl Ester, 1-Ethylallyl Acetate PubChem CID: 530323 IUPAC Name: pent-1-en-3-yl acetate SMILES: CCC(C=C)OC(=O)C