accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOHO

CAS RN 10502-44-0

IUPAC Name: 2-hydroxy-2-(4-methoxyphenyl)acetic acid
Synonyms: hydroxy(4-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: ITECRQOOEQWFPE-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results