accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10504-04-8

IUPAC Name: 2,3,5-trimethylfuran
Synonyms: 2,3,5-trimethylfuran
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: NJXZFRUNHWKHEC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results