CAS RN 105047-45-8
IUPAC Name:
(2S)-6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms:
(2S)-6-amino-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol):
368.43
Molecular Formula:
C21H24N2O4
InChi Key:
YRKFMPDOFHQWPI-IBGZPJMESA-N
SMILES:
NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Fmoc-Lys-OH, Advanced ChemTech
CAS: 105047-45-8 Molecular Formula: C21H24N2O4 Molecular Weight (g/mol): 368.43 MDL Number: MFCD00038539 InChI Key: YRKFMPDOFHQWPI-IBGZPJMESA-N IUPAC Name: (2S)-6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid SMILES: NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O