accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNCH3HCl

CAS RN 1050480-30-2

IUPAC Name: [(9-ethyl-9H-carbazol-3-yl)methyl](methyl)amine hydrochloride
Synonyms: [(9-ethylcarbazol-3-yl)methyl](methyl)amine hydrochloride
Molecular Weight (g/mol): 274.79
Molecular Formula: C16H19ClN2
InChi Key: IXWVUPURFWFRNE-UHFFFAOYSA-N
SMILES: Cl.CCN1C2=C(C=CC=C2)C2=C1C=CC(CNC)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results