accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONNH2NNHOHOHIOH3C

CAS RN 1050670-85-3

IUPAC Name: 2-({4-amino-5-[5-iodo-4-methoxy-2-(propan-2-yl)phenoxy]pyrimidin-2-yl}amino)propane-1,3-diol
Synonyms: 2-{[4-amino-5-(5-iodo-2-isopropyl-4-methoxyphenoxy)pyrimidin-2-yl]amino}propane-1,3-diol
Molecular Weight (g/mol): 474.3
Molecular Formula: C17H23IN4O4
InChi Key: PAYROHWFGZADBR-UHFFFAOYSA-N
SMILES: COC1=C(I)C=C(OC2=CN=C(NC(CO)CO)N=C2N)C(=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results