CAS RN 105125-00-6
IUPAC Name:
4-bromo-5-(thiophen-2-yl)-2,2'-bithiophene
Synonyms:
4-bromo-5-(thiophen-2-yl)-2,2'-bithiophene
Molecular Weight (g/mol):
327.27
Molecular Formula:
C12H7BrS3
InChi Key:
FGBHDLKMGUOJBA-UHFFFAOYSA-N
SMILES:
BrC1=C(SC(=C1)C1=CC=CS1)C1=CC=CS1
1
–
1
of
1
results
1
3'-Bromo-2,2':5',2″-terthiophene 97.0+%, TCI America™
CAS: 105125-00-6 Molecular Formula: C12H7BrS3 Molecular Weight (g/mol): 327.272 InChI Key: FGBHDLKMGUOJBA-UHFFFAOYSA-N Synonym: 3-Bromo-2,5-di(2-thienyl)thiophene PubChem CID: 184442 IUPAC Name: 3-bromo-2,5-dithiophen-2-ylthiophene SMILES: C1=CSC(=C1)C2=CC(=C(S2)C3=CC=CS3)Br