accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10516-71-9

IUPAC Name: 3-(3-methoxyphenyl)propanoic acid
Synonyms: 3-(3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: BJJQJLOZWBZEGA-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CCC(O)=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)