accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNCl

CAS RN 10517-21-2

IUPAC Name: 5-chloro-1H-indole-2-carboxylate
Synonyms: 5-chloroindole-2-carboxylate
Molecular Weight (g/mol): 194.59
Molecular Formula: C9H5ClNO2
InChi Key: FUQOTYRCMBZFOL-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)