accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10517-44-9

IUPAC Name: dihydrogen propane-1,3-diamine dichloride
Synonyms: dihydrogen α,ω-propanediamine dichloride
Molecular Weight (g/mol): 147.04
Molecular Formula: C3H12Cl2N2
InChi Key: HYOCSVGEQMCOGE-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCCN
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)