accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNOH2NOHO

CAS RN 10521-49-0

IUPAC Name: 2-(2-aminoacetamido)-3-methylbutanoic acid
Synonyms: L-valine, N-glycyl-
Molecular Weight (g/mol): 174.2
Molecular Formula: C7H14N2O3
InChi Key: STKYPAFSDFAEPH-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)CN)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results