accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHHN

CAS RN 105250-17-7

IUPAC Name: 2-amino-4-(hydroxymethyl)pyridin-1-ium
Synonyms: 2-amino-4-(hydroxymethyl)pyridin-1-ium
Molecular Weight (g/mol): 125.15
Molecular Formula: C6H9N2O
InChi Key: ZRJJXXDQIQFZBW-UHFFFAOYSA-O
SMILES: NC1=CC(CO)=CC=[NH+]1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results