accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10531-16-5

IUPAC Name: 1-(bromomethyl)-4-butylbenzene
Synonyms: 1-(bromomethyl)-4-butylbenzene
Molecular Weight (g/mol): 227.15
Molecular Formula: C11H15Br
InChi Key: XNQBGVFFKGZXJH-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(CBr)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results