accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONSH2N

CAS RN 105355-26-8

IUPAC Name: 2-amino-5-({4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl}methyl)-4,5-dihydro-1,3-thiazol-4-one
Synonyms: 2-amino-5-({4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl}methyl)-5H-1,3-thiazol-4-one
Molecular Weight (g/mol): 355.46
Molecular Formula: C19H21N3O2S
InChi Key: ABGJULHDDNEULW-UHFFFAOYNA-N
SMILES: CCC1=CN=C(CCOC2=CC=C(CC3SC(N)=NC3=O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results