accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1053655-56-3

IUPAC Name: 1-methyl-1H-indazole-4-carbaldehyde
Synonyms: 1-methylindazole-4-carbaldehyde
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: SONWSIAHVBTXPB-UHFFFAOYSA-N
SMILES: CN1N=CC2=C(C=O)C=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results