accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10538-51-9

IUPAC Name: (2E)-3-(2,5-dimethoxyphenyl)prop-2-enoate
Synonyms: (2E)-3-(2,5-dimethoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 207.21
Molecular Formula: C11H11O4
InChi Key: JPQWWJZORKTMIZ-ZZXKWVIFSA-M
SMILES: COC1=CC=C(OC)C(\C=C\C([O-])=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results