accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOHO

CAS RN 10541-83-0

IUPAC Name: 4-(methylamino)benzoic acid
Synonyms: N-methyl-4-aminobenzoate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: ZVIDMSBTYRSMAR-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)