accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1054483-78-1

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol
Molecular Weight (g/mol): 221.06
Molecular Formula: C11H16BNO3
InChi Key: WAUWXCUPDOXYKS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results