accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnH3CCH3NNH3C

CAS RN 105494-69-7

IUPAC Name: 1-methyl-2-(tributylstannyl)-1H-imidazole
Synonyms: 1-methyl-2-(tributylstannyl)imidazole
Molecular Weight (g/mol): 371.16
Molecular Formula: C16H32N2Sn
InChi Key: KFWFYOPKNSYTMV-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results