accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105496-31-9

IUPAC Name: (9H-fluoren-9-yl)methyl N-(2-hydroxyethyl)carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-(2-hydroxyethyl)carbamate
Molecular Weight (g/mol): 283.33
Molecular Formula: C17H17NO3
InChi Key: XLIFWDZVNRWYKV-UHFFFAOYSA-N
SMILES: OCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)