accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10553-14-7

IUPAC Name: 7-nitro-1H-indole-3-carbaldehyde
Synonyms: 7-nitro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 190.16
Molecular Formula: C9H6N2O3
InChi Key: ADGKBVRTGVODMM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2NC=C(C=O)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results