accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNH2H2NOHHClHCl

CAS RN 1055301-14-8

IUPAC Name: 4-[1,2-diamino-2-(4-hydroxyphenyl)ethyl]phenol dihydrochloride
Synonyms: 4-[1,2-diamino-2-(4-hydroxyphenyl)ethyl]phenol dihydrochloride
Molecular Weight (g/mol): 317.21
Molecular Formula: C14H18Cl2N2O2
InChi Key: WSCIOLVKHQOSNF-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)