accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1055301-16-0

IUPAC Name: 1,2-bis(2-chlorophenyl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(2-chlorophenyl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 354.1
Molecular Formula: C14H16Cl4N2
InChi Key: YXGVUHANCVCDJJ-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=CC=CC=C1Cl)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results