accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOIOH

CAS RN 105551-42-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 316.11
Molecular Formula: C7H9IO4S
InChi Key: WSUYPCBSGVAXNY-UHFFFAOYSA-N
SMILES: CS(=O)(=O)O[I](O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results