accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNClCl

CAS RN 10557-51-4

IUPAC Name: 5-(4-chlorophenyl)-2,3-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 5-(4-chlorophenyl)-2,3-diphenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 369.25
Molecular Formula: C19H14Cl2N4
InChi Key: CCQVUWNFRJMAMY-UHFFFAOYSA-M
SMILES: [Cl-].ClC1=CC=C(C=C1)C1=NN(C2=CC=CC=C2)[N+](=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results