CAS RN 10560-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,3-Diphenyl-5-(4-methoxyphenyl)tetrazolium Chloride 80.0+%, TCI America™
CAS: 10560-45-9 Molecular Formula: C20H17ClN4O MDL Number: MFCD00060032