accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3SOI

CAS RN 10561-14-5

IUPAC Name: [2-(benzoylsulfanyl)ethyl]trimethylazanium iodide
Synonyms: [2-(benzoylsulfanyl)ethyl]trimethylazanium iodide
Molecular Weight (g/mol): 351.25
Molecular Formula: C12H18INOS
InChi Key: PCGVSJGEHWZOBK-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)CCSC(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results