accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHHNNH2

CAS RN 10563-26-5

IUPAC Name: (3-aminopropyl)({2-[(3-aminopropyl)amino]ethyl})amine
Synonyms: 1,5,8,12-tetraazadodecane 3,2,3-tetramine
Molecular Weight (g/mol): 174.29
Molecular Formula: C8H22N4
InChi Key: RXFCIXRFAJRBSG-UHFFFAOYSA-N
SMILES: NCCCNCCNCCCN
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)