accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNBr

CAS RN 105655-01-4

IUPAC Name: 6-bromo-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 6-bromo-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: RWKBNMSHIJBNAO-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCCN2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results