accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105655-17-2

CH3OOBrNH2NOO

CAS RN 105655-17-2

IUPAC Name: methyl 4-amino-3-bromo-5-nitrobenzoate
Synonyms: methyl 4-amino-3-bromo-5-nitrobenzoate
Molecular Weight (g/mol): 275.06
Molecular Formula: C8H7BrN2O4
InChi Key: PRTLMEWWYBJZPN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(N)C(Br)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results