accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105669-53-2

IUPAC Name: N-[2-(dimethylamino)ethyl]-N-methylformamide
Synonyms: N-[2-(dimethylamino)ethyl]-N-methylformamide
Molecular Weight (g/mol): 130.19
Molecular Formula: C6H14N2O
InChi Key: CSLBJRKWKVBRSQ-UHFFFAOYSA-N
SMILES: CN(C)CCN(C)C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results