accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105679-22-9

OHNBr

CAS RN 105679-22-9

IUPAC Name: 7-bromo-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 7-bromo-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: JLZUUGCTPRPFKZ-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results