accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OHONH2CH3CH3

CAS RN 105743-53-1

IUPAC Name: (2S)-2-amino-2,4-dimethylpentanoic acid
Synonyms: 2-methylleucine
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: ARSWQPLPYROOBG-JLDDOWRYNA-N
SMILES: CC(C)C[C@](C)(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results