accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10575-25-4

IUPAC Name: 4-(diethylamino)but-2-yn-1-ol
Synonyms: 4-(diethylamino)but-2-yn-1-ol
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: ACGZBRWTWOZSFU-UHFFFAOYSA-N
SMILES: CCN(CC)CC#CCO
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)