accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NI

CAS RN 105752-11-2

IUPAC Name: 4-iodopyridin-3-amine
Synonyms: 4-iodopyridin-3-amine
Molecular Weight (g/mol): 220.01
Molecular Formula: C5H5IN2
InChi Key: ZJRSKTXMSIVNAU-UHFFFAOYSA-N
SMILES: NC1=CN=CC=C1I
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)