accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3NHO(E)

CAS RN 10576-12-2

IUPAC Name: (E)-(ethyl N-hydroxyethanimidate)
Synonyms: (E)-(ethyl N-hydroxyethanimidate)
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: QWKAVVNRCKPKNM-SNAWJCMRSA-N
SMILES: CCO\C(C)=N\O
Boiling Point 
  • (5)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)