accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1058-92-0

IUPAC Name: disodium (3Z)-3-[2-(5-chloro-2-hydroxyphenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (3Z)-3-[2-(5-chloro-2-hydroxyphenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 518.8
Molecular Formula: C16H9ClN2Na2O9S2
InChi Key: RAIRBURLMTXYPH-WTBAQZMLSA-L
SMILES: [Na+].[Na+].OC1=CC=C(Cl)C=C1N\N=C1\C(=O)C2=C(O)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results