accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH(R)(S)HOF

CAS RN 105812-81-5

IUPAC Name: (3S,4R)-4-(4-fluorophenyl)-3-(hydroxymethyl)-1-methylpiperidin-1-ium
Synonyms: (3S,4R)-4-(4-fluorophenyl)-3-(hydroxymethyl)-1-methylpiperidin-1-ium
Molecular Weight (g/mol): 224.3
Molecular Formula: C13H19FNO
InChi Key: CXRHUYYZISIIMT-AAEUAGOBSA-O
SMILES: C[NH+]1CC[C@H]([C@H](CO)C1)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results