accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOO

CAS RN 10595-06-9

IUPAC Name: 2-phenoxyethyl 2-methylprop-2-enoate
Synonyms: phenoxyethyl methacrylate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: CEXQWAAGPPNOQF-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCOC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results