accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CNO

CAS RN 10595-95-6

IUPAC Name: ethyl(methyl)nitrosoamine
Synonyms: N-methyl-N-nitrosoethamine
Molecular Weight (g/mol): 88.11
Molecular Formula: C3H8N2O
InChi Key: RTDCJKARQCRONF-UHFFFAOYSA-N
SMILES: CCN(C)N=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results