CAS RN 1059624-23-5
IUPAC Name:
1-methyl-1-(prop-2-en-1-yl)pyrrolidin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms:
1-methyl-1-(prop-2-en-1-yl)pyrrolidin-1-ium; bistriflylimide anion
Molecular Weight (g/mol):
406.36
Molecular Formula:
C10H16F6N2O4S2
InChi Key:
RXYDHDCXRMFQBC-UHFFFAOYSA-N
SMILES:
C[N+]1(CC=C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
1-Allyl-1-methylpyrrolidinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
CAS: 1059624-23-5 Molecular Formula: C10H16F6N2O4S2 Molecular Weight (g/mol): 406.358 InChI Key: RXYDHDCXRMFQBC-UHFFFAOYSA-N PubChem CID: 132274870 IUPAC Name: bis(trifluoromethylsulfonyl)azanide;1-methyl-1-prop-2-enylpyrrolidin-1-ium SMILES: C[N+]1(CCCC1)CC=C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F