accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105969-98-0

IUPAC Name: 2-hydroxy-5-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-hydroxy-5-nitroisoindole-1,3-dione
Molecular Weight (g/mol): 208.13
Molecular Formula: C8H4N2O5
InChi Key: MKACMVMZUIQKNY-UHFFFAOYSA-N
SMILES: ON1C(=O)C2=C(C=C(C=C2)[N+]([O-])=O)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results